Learn More
Invitrogen™ PIWIL1 Polyclonal Antibody

Description
A suggested positive control is HepG2 cell lysate. PA5-21051 can be used with blocking peptide PEP-1165.
Specifications
Specifications
| Antigen | PIWIL1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PIWIL1 |
| Gene Accession No. | Q96J94, Q9JMB7 |
| Gene Alias | CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 |
| Gene Symbols | PIWIL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.