Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL1 Polyclonal Antibody
PIWIL1 Polyclonal Antibody
Supplier: Thermo Scientific PA596072
Description
PIWIL1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
PIWIL1 | |
Polyclonal | |
Unconjugated | |
PIWIL1 | |
CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
363912, 57749, 9271 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96J94, Q9JMB7 | |
PIWIL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of mouse PIWIL1 (NP_0672861). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction