Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241126
Description
PIWIL2 Polyclonal specifically detects PIWIL2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PIWIL2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
PIWIL2 | |
Antibody was raised against a 14 amino acid synthetic peptide near the center of human PIWI-L2. The immunogen is located within amino acids 460 - 510 of PIWI-L2. Amino Acid Squence: LIHRPSERQDNHGM | |
0.1 mg | |
Primary | |
Human | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
55124 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction