Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIWIL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568430
Description
Predicted to react with Mouse samples.
The piwi gene of Drosophila belongs to a novel class of evolutionarily conserved genes (the piwi or Argonaute family). The Piwi family is required for germ and stem cell development in invertebrates, and two Piwi members, MIWI and MILI (Miwi like, Piwil2, Piwi-like protein 2), are essential for spermatogenesis in mouse. MIWI and MILI partner with Piwi-interacting RNAs (piRNAs) which are 26-31 nucleotides in length clearly distinct from the 21-23 nucleotides of microRNAs (miRNAs) or short interfering RNAs (siRNAs). A search for murine small RNAs that might program Piwi proteins for transposon suppression revealed developmentally regulated piRNA loci, some of which resemble transposon master control loci of Drosophila. MILI might be programmed by distinct piRNA populations at different stages of germ cell development, suggesting MILI plays an important role in meiotic prophase progression.
Specifications
PIWIL2 | |
Polyclonal | |
Unconjugated | |
Piwil2 | |
80kDa PIWIL2 short isoform; cancer/testis antigen 80; CT80; HILI; mili; Miwi like; Miwi likw; piwi like homolog 2; piwi like RNA-mediated gene silencing 2; PIWIL1L; Piwil2; PIWI-L2; piwil2-like protein; piwi-like 2; piwi-like homolog 2; piwi-like protein 2; piwi-like RNA-mediated gene silencing 2; testicular tissue protein Li 141 | |
Rabbit | |
affinity chromatography | |
RUO | |
55124, 57746 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8CDG1, Q8TC59 | |
Piwil2 | |
Synthesized peptide derived from human PIWIL2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction