Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176490
Description
PIWIL3 Polyclonal specifically detects PIWIL3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PIWIL3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
HIWI3, piwi-like 3 (Drosophila), piwi-like protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
440822 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
NP_001008496 | |
PIWIL3 | |
Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PIWI-L3. The immunogen is located within amino acids 130 - 180 of PIWI-L3. Amino Acid Squence: YKYNVDYKPDIEDGNLRT | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction