Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIWIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521052
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521052 Supplier Invitrogen™ Supplier No. PA521052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-21052 can be used with blocking peptide PEP-1166.

PIWI-L3 is a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that are crucial for the biogenesis and function of small non-coding RNAs and play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. Recent studies have shown that overexpression of PIWI-L3 (as well as other PIWI-like proteins) are potential biomarkers for astrocytic glioma, meningioma, and other cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIWIL3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PIWIL3
Gene Accession No. Q7Z3Z3
Gene Alias HIWI3; piwi like RNA-mediated gene silencing 3; PIWIL3; piwi-like 3; piwi-like protein 3; piwi-like RNA-mediated gene silencing 3
Gene Symbols PIWIL3
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human PIWI-L3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440822
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.