Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIWIL4 Monoclonal Antibody (10G9B11)
GREENER_CHOICE

Catalog No. PIMA517151
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517151 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517151 Supplier Invitrogen™ Supplier No. MA517151
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17151 targets PIWIL4 in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human samples. The MA5-17151 immunogen is purified recombinant fragment of human PIWIL4 (amino acids: 304-434) expressed in E. Coli. MA5-17151 detects PIWIL4 which has a predicted molecular weight of approximately 96.6kDa.

PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIWIL4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10G9B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PIWIL4
Gene Accession No. Q7Z3Z4
Gene Alias 9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P
Gene Symbols PIWIL4
Host Species Mouse
Immunogen Purified recombinant fragment of human PIWIL4 (amino acids: 304-434) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.