Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIWIL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531448 Supplier Invitrogen™ Supplier No. PA531448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PIWIL4-transfected 293T. Predicted reactivity: Rat (80%), Pig (85%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIWIL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PIWIL4
Gene Accession No. Q7Z3Z4
Gene Alias 9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P
Gene Symbols PIWIL4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PIWIL4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.