Learn More
Invitrogen™ PJA Polyclonal Antibody

Description
Immunogen sequence: LPARPSRAPV SICGGGENTS KSAEEPVVRP KIRNLASPNC VKPKIFFDTD DDDDMPHSTS RWRDTANDNE GHSDGLARRG RGESSSGYPE PKYPEDKREA RSDQVKP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 60%.
Specifications
Specifications
| Antigen | PJA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PJA1 |
| Gene Accession No. | Q8NG27 |
| Gene Alias | E3 ubiquitin-protein ligase Praja-1; Pja1; praja 1; praja ring finger 1, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 1; Praja1; praja-1; praja1, RING-H2 motif containing; RING finger protein 70; RING-type E3 ubiquitin transferase Praja-1; RNF70 |
| Gene Symbols | PJA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.