Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PJA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572845
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572845 Supplier Invitrogen™ Supplier No. PA572845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PJA Polyclonal Antibody for Western Blot, IHC (P), ELISA

Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome. May be involved in protein sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PJA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PJA1
Gene Accession No. O55176, Q8NG27
Gene Alias E3 ubiquitin-protein ligase Praja-1; Pja1; praja 1; praja ring finger 1, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 1; Praja1; praja-1; praja1, RING-H2 motif containing; RING finger protein 70; RING-type E3 ubiquitin transferase Praja-1; RNF70
Gene Symbols PJA1
Host Species Rabbit
Immunogen PJA1 antibody for a 19 amino acid peptide near the amino terminus of human PJA1. The immunogen is within amino acids 50 - 100 of PJA1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18744, 64219, 683077
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.