Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PJA2 Polyclonal Antibody

Catalog No. PIPA535950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535950 Supplier Invitrogen™ Supplier No. PA535950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Human (99%), Mouse (99%), Rat (99%), Bovine (99%).

Has E2-dependent E3 ubiquitin-protein ligase activity. Responsible for ubiquitination of cAMP-dependent protein kinase type I and type II-alpha/beta regulatory subunits and for targeting them for proteasomal degradation. Essential for PKA-mediated long-term memory processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PJA2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PJA2
Gene Accession No. O43164, Q80U04
Gene Alias AI447901; AL022700; E3 ubiquitin-protein ligase Praja2; E3 ubiquitin-protein ligase Praja-2; Kiaa0438; mKIAA0438; Neurodap1; neurodegeneration associated protein 1; Pja2; praja 2, RING-H2 motif containing; praja ring finger 2, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 2; praja2; praja-2; protein carrying the RING-H2 sequence motif; RING finger protein 131; RING-type E3 ubiquitin transferase Praja-2; RNF131
Gene Symbols PJA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PJA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224938, 9867
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.