Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PJA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566096
Description
Immunogen sequence: DEMVSTKQQNN TSQERQTEHS PEDAACGPGH ICSEQNTNDR EKNHGSSPEQ VVRPKVRKLI SSSQVDQETG FNRHEAKQRS VQRW Highest antigen sequence identity to the following orthologs - mouse 73%, rat 72%.
Has E2-dependent E3 ubiquitin-protein ligase activity. Responsible for ubiquitination of cAMP-dependent protein kinase type I and type II-alpha/beta regulatory subunits and for targeting them for proteasomal degradation. Essential for PKA-mediated long-term memory processes.
Specifications
PJA2 | |
Polyclonal | |
Unconjugated | |
PJA2 | |
AI447901; AL022700; E3 ubiquitin-protein ligase Praja2; E3 ubiquitin-protein ligase Praja-2; Kiaa0438; mKIAA0438; Neurodap1; neurodegeneration associated protein 1; Pja2; praja 2, RING-H2 motif containing; praja ring finger 2, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 2; praja2; praja-2; protein carrying the RING-H2 sequence motif; RING finger protein 131; RING-type E3 ubiquitin transferase Praja-2; RNF131 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9867 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43164 | |
PJA2 | |
Recombinant Human PJA2. Recombinant protein control fragment (Product #RP-105081). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction