Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PJA2/Praja2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25690125UL
Description
PJA2/Praja2 Polyclonal specifically detects PJA2/Praja2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PJA2/Praja2 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
E3 ubiquitin-protein ligase Praja-2, EC 6.3.2, EC 6.3.2.-, KIAA0438praja2, Neurodap1, praja ring finger 2, Praja2, RING finger protein 131praja 2, RING-H2 motif containing, RNF131 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
PJA2 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DEMVSTKQQNNTSQERQTEHSPEDAACGPGHICSEQNTNDREKNHGSSPEQVVRPKVRKLISSSQVDQETGFNRHEAKQRSVQRW | |
25 μL | |
Zinc Finger | |
9867 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction