Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PJA2/Praja2 Rabbit anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
SDP

Catalog No. NB093134 Shop All Novus Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093134 20 μL
NB093133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093134 Supplier R&D Systems Supplier No. NBP30373020UL

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

PJA2/Praja2 Polyclonal specifically detects PJA2/Praja2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PJA2/Praja2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias E3 ubiquitin-protein ligase Praja-2, EC 6.3.2, EC 6.3.2.-, KIAA0438praja2, Neurodap1, praja ring finger 2, Praja2, RING finger protein 131praja 2, RING-H2 motif containing, RNF131
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 439-708 of human PJA2/Praja2 (NP_055634.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9867
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.