Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PK2L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116235
Description
PK2L1 Polyclonal Antibody for Western Blot
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined.
Specifications
PK2L1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9P0L9 | |
Pkd2l1 | |
KLH conjugated synthetic peptide between 37-65 amino acids from the N-terminal region of human PKD2L1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Pkd2l1 | |
B830002B15; BC046386; PCL; PK2L1; PKD2L; PKD2L1; Pkdl; polycystic kidney disease 2-like 1; polycystic kidney disease 2-like 1 protein; polycystin 2 like 1, transient receptor potential cation channel; polycystin-2 homolog; Polycystin-2L1; polycystin-L; Polycystin-L1; transient receptor potential cation channel, subfamily P, member 3; TRPP3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
9033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction