Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PK2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144981 Supplier Invitrogen™ Supplier No. PA5144981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNKTLLRLRL RKERVSDVQK VLQGGEQEIQ FEDFTNTLRE LGHAEHEITE LTATFTKFDR DGNRILDEKE QE Antigen sequence identity: Mouse 82%; Rat 51%.

This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PK2L1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pkd2l1
Gene Accession No. Q9P0L9
Gene Alias B830002B15; BC046386; PCL; PK2L1; PKD2L; PKD2L1; Pkdl; polycystic kidney disease 2-like 1; polycystic kidney disease 2-like 1 protein; polycystin 2 like 1, transient receptor potential cation channel; polycystin-2 homolog; Polycystin-2L1; polycystin-L; Polycystin-L1; transient receptor potential cation channel, subfamily P, member 3; TRPP3
Gene Symbols Pkd2l1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 588-659. Recombinant protein control fragment (Product #RP-110142).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.