Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKA alpha/beta/gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567660 Supplier Invitrogen™ Supplier No. PA567660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKA alpha/beta/gamma Polyclonal Antibody for Western Blot, ICC/IF

Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKA alpha/beta/gamma
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKACA
Gene Accession No. P17252, P22612, P22694
Gene Alias BDPLT19; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit beta; cAMP-dependent protein kinase catalytic subunit gamma; KAPG; PKA C-alpha; PKA C-beta; PKA C-gamma; PKACA; PKACB; PKACg; PPNAD4; PRKACA; PRKACB; PRKACG; protein kinase A catalytic subunit; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit alpha; protein kinase cAMP-activated catalytic subunit beta; protein kinase cAMP-activated catalytic subunit gamma; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; protein kinase, cAMP-dependent, catalytic, beta; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase
Gene Symbols PRKACA, PRKACB, PRKACG
Host Species Rabbit
Immunogen Synthesized peptide derived from human PKA alpha/beta/gamma.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5566, 5567, 5568
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.