Learn More
Invitrogen™ PKA alpha Polyclonal Antibody

Description
Recommended positive controls: HeLaS3, NIH-3T3, C2C12. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (94%), Dog (99%), Rabbit (97%), Sheep (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PKA alpha |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PRKACA |
| Gene Accession No. | P05132, P17252 |
| Gene Alias | alternatively spliced; C(s); Calpha; C-alpha-2; C-alpha-S; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; cAMP-dependent protein kinase catalytic subunit Calpha1; Cs-PKA; MGC102831; MGC48865; PKA; PKA C-alpha; PKA catalytic subunit alpha; pkac; Pkaca; PKCA1; PKCD; PPNAD4; PRKACA; protein kinase; protein kinase A; protein kinase A alpha; protein kinase A catalytic subunit; protein kinase cAMP-activated catalytic subunit alpha; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; sperm cAMP-dependent protein kinase catalytic subunit; sperm cAMP-dependent protein kinase catalytic subunit Cs |
| Gene Symbols | PRKACA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.