Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKA beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117489 Supplier Invitrogen™ Supplier No. PA5117489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family (the PKA catalytic subunit consist of three gene products: C-alpha, C-beta, and C-gamma) and has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKA beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRKACB
Gene Accession No. P22694, P68181, P68182
Gene Alias cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit beta; PKA C-beta; Pkacb; PRKACB; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit beta; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, beta
Gene Symbols PRKACB
Host Species Rabbit
Immunogen Recombinant Human cAMP-dependent protein kinase catalytic subunit beta protein (2-53aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18749, 293508, 5567
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.