Learn More
Promega PKA Kinase Enzyme System, 2,500 units
Catalog No. PRV4246 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
2500 Units
Catalog No. Quantity
PRV4246 2500 Units
1 options
Catalog No. PRV4246 Supplier Promega Supplier No. V4246
Recombinant bovine cAMP-Dependent Protein Kinase (PKA), Catalytic Subunit, is expressed and purified from E. coli. PKA does not require cAMP for activity, and the gene sequence can be found at accession number NM_174584.
  • Easily Screen and Profile PKA Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 2500 Units
Content And Storage <-65°C