Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKA R2, p Ser99 Antibody (SR1905), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32151150UL
Description
PKA R2 Monoclonal antibody specifically detects PKA R2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
PKA R2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1905 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit | |
A synthesized peptide derived from human phospho-PKA R2 [Ser99] (Uniprot #: P13861) | |
50 μg | |
Phospho Specific, Signal Transduction | |
5576 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction