Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC alpha Antibody (133), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308229AF647
Description
PKC alpha Monoclonal antibody specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
PKC alpha | |
Monoclonal | |
Alexa Fluor 647 | |
AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
Full-length protein corresponding to PKC alpha. Recognizes sequence PQFVHPILQSAV at the C terminus at PKCalpha. (Uniprot: P17252) | |
100 μg | |
Cancer, Cellular Markers, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
5578 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin) | |
133 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction