Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC alpha Antibody (G13-R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP1301210.05ML
Description
PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
PKC alpha | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1:5000, ELISA 1:20000-1:50000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
P17252 | |
PRKCA | |
Peptide derived from the human PKC-alpha sequence | |
0.05 mL | |
Cancer, Cellular Markers, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
5578 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
G13-R | |
Unconjugated | |
20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction