Learn More
Promega PKC alpha Kinase Enzyme System, 1mg
Catalog No. PRV3382 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3382 1 mg
1 options
Catalog No. PRV3382 Supplier Promega Supplier No. V3382
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKC alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC alpha is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C