Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11086972
Description
PKC beta Polyclonal specifically detects PKC beta in Human samples. It is validated for Western Blot, ELISA.Specifications
PKC beta | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
EC 2.7.11, EC 2.7.11.13, MGC41878, PKC-B, PKC-beta, PKCBPRKCB2, PRKCB1protein kinase C beta 1, protein kinase C beta type, protein kinase C, beta, protein kinase C, beta 1, protein kinase C, beta 1 polypeptide | |
Rabbit | |
76.87 kDa | |
0.1 mg | |
mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
5579 | |
Human, Primate | |
IgG |
Western Blot, ELISA | |
1 mg/ml | |
Western Blot 1:1000-1:2000, ELISA 1:75000-1:80000 | |
P05771 | |
PRKCB | |
This affinity purified PKC beta Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human PKC beta. (Uniprot: P05771) | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for human PKC beta protein. A BLAST analysis was used to suggest cross-reactivity with PKC beta from human and monkey sources based on a 100% identity with the immunizing sequence. Expect partial cross reactivity to PKC beta from rabbit and bovine (93% identity) and rat and mouse (87% identity) sources. Cross-reactivity with PKC beta from other sources has not been determined. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction