Learn More
Promega PKC beta I Kinase Enzyme System, 1mg
Catalog No. PRV6595 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6595 1 mg
1 options
Catalog No. PRV6595 Supplier Promega Supplier No. V6595
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKCbeta I was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKCβI is a member of the PKC family (phospholipid-dependent serine/threonine kinase) and is highly related to PKCβ II.
  • Easily Screen and Profile PKCβI Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C