Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC delta Antibody (M242), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323307
Description
PKC delta Monoclonal antibody specifically detects PKC delta in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PKC delta | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
M242 | |
Western Blot 1:250, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 2.7.11, EC 2.7.11.13, MAY1, MGC49908, nPKC-delta, PKCD, PKCy, protein kinase C delta type, protein kinase C delta VIII, protein kinase C, delta | |
Clone (M242) was generated from a recombinant human protein that included amino acids residues in the N-terminal region. This sequence is conserved in rat and mouse PKC delta, and has low homology to other PKC family members. [UniProt# Q05655] | |
0.1 mL | |
Phospho Specific, Protein Kinase, Signal Transduction | |
5580 | |
Store at -20°C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction