Learn More
Promega PKC delta Kinase Enzyme System, 1mg
Catalog No. PRV3402 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3402 1 mg
1 options
Catalog No. PRV3402 Supplier Promega Supplier No. V3402
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKC delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C delta is a member of the protein kinase C (PKC) family of serine-threonine kinases.
  • Easily Screen and Profile PKCδ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C