Learn More
PKC-delta Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302448AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.Specifications
PKC-delta | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q05655 | |
PRKCD | |
Between 626 and 676 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRKCD | |
ALPS3, CVID9, MAY1, nPKC-delta, PKCD, protein kinase C delta VIII, protein kinase C, delta, tyrosine-protein kinase PRKCD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5580 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.