Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC epsilon Antibody (OTI4G8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273438AF750
Description
PKC epsilon Monoclonal specifically detects PKC epsilon in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PKC epsilon | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
PRKCE | |
Full length recombinant protein of human PKC epsilon (NP_005391) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4G8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCEMGC125657, protein kinase C epsilon type, protein kinase C, epsilon | |
Mouse | |
0.1 mL | |
5581 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction