Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKC eta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582908 Supplier Invitrogen™ Supplier No. PA582908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKEIDWAQLN HRQIEPPFRP RIKSREDVSN FDPDFIKEEP VLTPIDEGHL PMINQDEFRN F Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 97%.

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC eta
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRKCH
Gene Accession No. P24723
Gene Alias LOW QUALITY PROTEIN: protein kinase C eta type; MGC26269; MGC5363; nPKC-eta; PKC; Pkch; PKCL; PKC-L; PRKCH; PRKCL; protein kinase C eta; protein kinase C eta type; protein kinase C eta type pseudogene; protein kinase C theta; protein kinase C, eta; protein kinase C-eta; Unknown (protein for MGC:140784)
Gene Symbols PRKCH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRKCH. Recombinant protein control fragment (Product #RP-95224).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.