Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKC eta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119589
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications
PKC eta | |
Polyclonal | |
Unconjugated | |
PRKCH | |
LOW QUALITY PROTEIN: protein kinase C eta type; MGC26269; MGC5363; nPKC-eta; PKC; Pkch; PKCL; PKC-L; PRKCH; PRKCL; protein kinase C eta; protein kinase C eta type; protein kinase C eta type pseudogene; protein kinase C theta; protein kinase C, eta; protein kinase C-eta; Unknown (protein for MGC:140784) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18755 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.07 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P23298 | |
PRKCH | |
Recombinant protein encompassing a sequence within the C-terminus region of mouse PKC eta. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction