Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC iota Antibody (PRKCI/4911), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314155H
This item is not returnable.
View return policy
Description
PKC iota Monoclonal antibody specifically detects PKC iota in Human, Canine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PKC iota | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine, Feline | |
Antibody | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PRKCI/4911 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota | |
Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743) | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
5584 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction