Learn More
Promega PKC iota Kinase Enzyme System, 1mg
Catalog No. PRV3752 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3752 1 mg
1 options
Catalog No. PRV3752 Supplier Promega Supplier No. V3752
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKC iota was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C iota is a member of the protein kinase C family of serine-threonine kinases.
  • Easily Screen and Profile PKCι Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C