Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKC lambda/iota Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579872
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579872 Supplier Invitrogen™ Supplier No. PA579872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SHG whole cell, A549 whole cell, U87 whole cell, 293T whole cell, HELA whole cell, JURKAT whole cell. IHC: Mouse Brain Tissue, Rat Brain Tissue, Human Lung Cancer Tissue.

The PKC family of serine/threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC lambda/iota
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene PRKCI
Gene Accession No. F1M7Y5, P41743, Q62074
Gene Alias 2310021H13Rik; AI427505; aPKClambda; aPKC-lambda/iota; atypical protein kinase C-lambda/iota; DXS1179E; hypothetical protein; mKIAA4165; nPKC-iota; Pkci; Pkcl; PKClambda; PRKCI; Prkcl; PRKC-lambda/iota; protein kinase C iota; protein kinase C iota type; protein kinase C, iota; protein kinase C, lambda
Gene Symbols PRKCI
Host Species Rabbit
Immunogen E.coli-derived human PKC iota recombinant protein (Position: D21-Q214).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18759, 5584, 84006
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.