Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC lambda/iota Polyclonal Antibody, Invitrogen™

Catalog No. 400600Z
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-060-0Z 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-060-0Z Supplier Invitrogen Supplier No. 400600Z

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

PKC lambda/iota Polyclonal antibody specifically detects PKC lambda/iota in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC lambda/iota
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PRKCI
Gene Accession No. F1M7Y5, P41743, Q62074
Gene Alias 2310021H13Rik; AI427505; aPKClambda; aPKC-lambda/iota; atypical protein kinase C-lambda/iota; DXS1179E; hypothetical protein; mKIAA4165; nPKC-iota; Pkci; Pkcl; PKClambda; PRKCI; Prkcl; PRKC-lambda/iota; protein kinase C iota; protein kinase C iota type; protein kinase C, iota; protein kinase C, lambda
Gene Symbols PRKCI
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of mouse, rat and human PKC-iota.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18759, 5584, 84006
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.