Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKC mu Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144355
Description
PKC mu Polyclonal Antibody for ICC/IF, ELISA
The PKC family of serine/threonine kinases, including PRKCN (PKD3), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC mu activation is dependent on the phosphorylation of two activation loop sites, Ser744 and Ser748, via a PKC-dependent signaling pathway.
Specifications
PKC mu | |
Polyclonal | |
Unconjugated | |
Prkd3 | |
4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3 | |
Rabbit | |
Protein G | |
RUO | |
23683, 25865, 5587 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
2.78 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
O94806, Q15139, Q9BZL6 | |
PRKD1, Prkd2, Prkd3 | |
Recombinant Human Serine/threonine-protein kinase D1 protein (674-904AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction