Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKC mu Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115765 Supplier Invitrogen™ Supplier No. PA5115765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PKD1/PKC mu.

The PKC family of serine/threonine kinases, including PRKCN (PKD3), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC mu activation is dependent on the phosphorylation of two activation loop sites, Ser744 and Ser748, via a PKC-dependent signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC mu
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Prkd3
Gene Accession No. O94806, Q15139, Q5XIS9, Q62101, Q8BZ03, Q8K1Y2, Q9BZL6, Q9WTQ1
Gene Alias 4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3
Gene Symbols PRKD1, Prkd2, Prkd3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRKD1(Accession Q15139), corresponding to amino acid residues R439-P489.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101540, 18760, 23683, 25865, 292658, 313834, 5587, 75292, 85421
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.