Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC theta Antibody (453416), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4368P
Description
PKC theta Monoclonal antibody specifically detects PKC theta in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
PKC theta | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF | |
453416 | |
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
EC 2.7.11, EC 2.7.11.13, MGC126514, MGC141919, nPKC-theta, PRKCT, protein kinase C theta type, protein kinase C, theta | |
E. coli-derived recombinant human PKC theta , His580-Ser706, Accession # Q04759 | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
5588 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction