Learn More
Promega PKC theta Kinase Enzyme System, 1mg
Catalog No. PRV6475 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6475 1 mg
1 options
Catalog No. PRV6475 Supplier Promega Supplier No. V6475
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKC theta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein Kinase C theta (PKC theta) is an important component in the intracellular signaling cascade.
  • Easily Screen and Profile PKCθ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C