Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC zeta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | PKC zeta |
---|---|
Concentration | 1.0 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017876
![]() |
Novus Biologicals
NB100821470.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB10082147
![]() |
Novus Biologicals
NB10082147 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
PKC zeta Polyclonal specifically detects PKC zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PKC zeta | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Rat, Human, Mouse | |
EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2PKC-ZETA, protein kinase C zeta type, protein kinase C, zeta | |
PRKCZ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Apoptosis, Diabetes Research | |
Q05513 | |
5590 | |
IgG | |
Affinity Purified | |
78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title