Learn More
Promega PKC zeta Kinase Enzyme System, 1mg
Catalog No. PRV2782 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2782 1 mg
1 options
Catalog No. PRV2782 Supplier Promega Supplier No. V2782
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PKC zeta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC zeta is an atypical isoform of the PKC family.
  • Easily Screen and Profile PKCζ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C