Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC zeta, p Thr560 Antibody (SR2127), Novus Biologicals™
SDP

Catalog No. NB283519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB283519 50 μg
NB284770 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB283519 Supplier Novus Biologicals Supplier No. NBP32155350UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PKC zeta Monoclonal antibody specifically detects PKC zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen PKC zeta
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR2127
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2PKC-ZETA, protein kinase C zeta type, protein kinase C, zeta
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-PKC zeta [Thr560] (Uniprot #: Q05513)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Diabetes Research, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5590
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.