Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC zeta Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501726663 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6663 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6663 Supplier Proteintech Group Inc Supplier No. 108601AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a constitutive kinase activity which is independent of calcium, and PKC activators, phosphatidylserine and diacylglycerol. Furthermore, it is insensitive to PKC inhibitors and cannot be activated by phorbol ester. The structural and biochemical properties indicate that the zeta subspecies is related to, but distinct from other isoenzymes of PKC.

Specifications

Antigen PKC zeta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PRKCZ
Gene Accession No. Q02956, Q05513
Gene Alias PKC ZETA, PKC2, PRKCZ, Protein kinase C zeta type, protein kinase C, zeta
Gene Symbols Prkcz
Host Species Rabbit
Immunogen PKC zeta Fusion Protein Ag1310
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18762, 5590
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.