Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PKCθ/δ Inhibitor

Catalog No. 5396495mg Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg

A cell-permeable pyridinecarbonitrile compound that acts as a potent, ATP-competitive (Ki = 79 nM, using PKCθ KD in coupled PK/LDH assays), and isozyme-selective PKCθ inhibitor (IC50 = 70nM) with much reduced potency against other novel PKCs (IC50

A cell-permeable pyridinecarbonitrile compound that acts as a potent, ATP-competitive (Ki = 79 nM, using PKCθ KD in coupled PK/LDH assays), and isozyme-selective PKCθ inhibitor (IC50 = 70nM) with much reduced potency against other novel PKCs (IC50 = 0.35, 2.33, and 16.35μM against δ, ε, and η isoforms, respectively) and little or no activity against the conventional PKCβ and atypical PKCζ (IC50 >50μM) as well as a panel of 18 other commonly studied cellular kinases (IC50 >10μM). Shown to preferentially inhibit IL-2 production from anti-CD3- and anti-CD28-activated T cells derived from wild-type over PKCθ-knockout mice (IC50 = 58 and 231 nM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Description 95% by HPLC
Molecular Formula C22H18N4O2
Formula Weight 370.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.