Learn More
Invitrogen™ PKD1L2 Polyclonal Antibody

Description
Immunogen sequence: MGEDSPVAMF SWYLDNTPTE QAEPLLDACR LRGFWPRSLT LLQSNTSTLL LNSSFLQSRG EVIRIRATAL TRHAYGEDTY VISTVPPREV PACTIAPEEG TVLTSFAIFC NASTALGPLE FCFCLESGSC LHCGPEPALP SVYLPLGEEN NDFVLTVVIS ATNRAGDTQQ TQAMAKVALG DTCVEDVAFQ AAVSEKIPTA LQGEGGPEQL LQLAKAVSSM LNQEHESQGS GQSLSIDVRQ KVREHVLGSL SAVTTGLEDV QRVQELAEVL REVTCRSKEL TPSAQGSCMG DSWEGAPPAA HVSHAR; Positive Samples: Mouse lung; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | PKD1L2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Pkd1l2 |
| Gene Accession No. | Q7TN88 |
| Gene Alias | 1700126L06Rik; KIAA1879; PC1L2; PC1-like 2 protein; PKD1L2; polycystic kidney disease 1 like 2; polycystic kidney disease 1-like 2; polycystic kidney disease protein 1-like 2; polycystin 1 like 2 (gene/pseudogene); polycystin-1L2 |
| Gene Symbols | Pkd1l2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.