Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKD1L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982381
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2381 Supplier Bioss Supplier No. BS12691R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the polycystin protein family. The encoded protein contains 11 transmembrane domains, a latrophilin/CL-1-like GPCR proteolytic site (GPS) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. This protein may function as a component of cation channel pores. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen PKD1L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pkd1l2
Gene Accession No. Q7Z442
Gene Alias 1700126L06Rik; KIAA1879; PC1L2; PC1-like 2 protein; Pkd1l2; polycystic kidney disease 1 like 2; polycystic kidney disease 1-like 2; Polycystic kidney disease protein 1-like 2; polycystin 1 like 2 (gene/pseudogene); Polycystin-1L2
Gene Symbols Pkd1l2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PKD1L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114780
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.