Learn More
Promega PKD2 Kinase Enzyme System, 10microg
Catalog No. PRV4042 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4042 10 μg
1 options
Catalog No. PRV4042 Supplier Promega Supplier No. V4042
Recombinant full-length human PKD2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKD2 is a novel phorbol ester- and growth factor-stimulated serine/threonine kinase.
  • Easily Screen and Profile PKD2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C