Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKD2L2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297274PE
Description
PKD2L2 Polyclonal antibody specifically detects PKD2L2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PKD2L2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
polycystic kidney disease 2-like 2, polycystic kidney disease 2-like 2 protein, polycystin-L2 | |
Produced in rabbits immunized with E. coli-derived Human PKD2L2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
27039 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction