Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKDREJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556982 Supplier Invitrogen™ Supplier No. PA556982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPVYEEPSDE VEAMTYLCRK LRTMFSFLTS QSKAKDEPEF FIDMLYGQPE KNSHRYLGLK TRNINGKKMV YL Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.

This intronless gene encodes a member of the polycystin protein family. The encoded protein contains 11 transmembrane domains, a receptor for egg jelly (REJ) domain, a G-protein-coupled receptor proteolytic site (GPS) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. This protein may play a role in human reproduction. Alternative splice variants have been described but their biological natures have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKDREJ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PKDREJ
Gene Accession No. Q9NTG1
Gene Alias PKD and REJ homolog; PKDREJ; polycystic kidney disease (polycystin) and REJ homolog (sperm receptor for egg jelly homolog, sea urchin); polycystic kidney disease and receptor for egg jelly-related protein; polycystin (PKD) family receptor for egg jelly
Gene Symbols PKDREJ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PKDREJ. Recombinant protein control fragment (Product #RP-95367).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.